Assay Details
Gene Symbol: gem | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299342.1 | NP_001286271.1 | 3 - 4 | 400 | 84 | |
NM_136712.4 | NP_610556.1 | 10 - 11 | 5219 | 84 | ||
NM_165748.3 | NP_724893.3 | 10 - 11 | 3561 | 84 | ||
NM_165750.4 | NP_724895.1 | 10 - 11 | 3977 | 84 | ||
GenBank mRNA | AY075312.1 | - | 10 - 11 | 4127 | 84 |
Gene Symbol: CG46319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347798.1 | NP_001334715.1 | 10 - 11 | 3977 | 84 | |
NM_001347799.1 | NP_001334716.1 | 10 - 11 | 5219 | 84 | ||
NM_001347800.1 | NP_001334717.1 | 10 - 11 | 3561 | 84 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
