Assay Details
Gene Symbol: gem | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299342.1 | NP_001286271.1 | 4 - 5 | 471 | 59 | |
NM_136712.4 | NP_610556.1 | 11 - 12 | 5290 | 59 | ||
NM_165748.3 | NP_724893.3 | 11 - 12 | 3632 | 59 | ||
NM_165750.4 | NP_724895.1 | 11 - 12 | 4048 | 59 | ||
GenBank mRNA | AY075312.1 | - | 11 - 12 | 4198 | 59 |
Gene Symbol: CG46319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347798.1 | NP_001334715.1 | 11 - 12 | 4048 | 59 | |
NM_001347799.1 | NP_001334716.1 | 11 - 12 | 5290 | 59 | ||
NM_001347800.1 | NP_001334717.1 | 11 - 12 | 3632 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
