Assay Details
Gene Symbol: gem | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299342.1 | NP_001286271.1 | 2 - 3 | 283 | 74 | |
NM_136712.4 | NP_610556.1 | 9 - 10 | 5102 | 74 | ||
NM_165748.3 | NP_724893.3 | 9 - 10 | 3444 | 74 | ||
NM_165750.4 | NP_724895.1 | 9 - 10 | 3860 | 74 | ||
GenBank mRNA | AY075312.1 | - | 9 - 10 | 4010 | 74 |
Gene Symbol: CG46319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347798.1 | NP_001334715.1 | 9 - 10 | 3860 | 74 | |
NM_001347799.1 | NP_001334716.1 | 9 - 10 | 5102 | 74 | ||
NM_001347800.1 | NP_001334717.1 | 9 - 10 | 3444 | 74 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
