Assay Details
Gene Symbol: sqh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297999.1 | NP_001284928.1 | 2 - 3 | 641 | 69 | |
NM_001298000.1 | NP_001284929.1 | 2 - 3 | 951 | 69 | ||
NM_001298001.1 | NP_001284930.1 | 2 - 3 | 645 | 69 | ||
NM_001298002.1 | NP_001284931.1 | 1 - 2 | 572 | 69 | ||
NM_078502.3 | NP_511057.1 | 2 - 3 | 665 | 69 | ||
GenBank mRNA | AY122159.1 | - | 2 - 3 | 636 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
