Assay Details
| Gene Symbol: RpL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298015.1 | NP_001284944.1 | 3 - 4 | 226 | 73 | |
| NM_001298016.1 | NP_001284945.1 | 3 - 4 | 226 | 73 | ||
| NM_078508.6 | NP_511063.1 | 3 - 4 | 315 | 73 | ||
| NM_167073.3 | NP_727094.1 | 4 - 5 | 339 | 73 | ||
| NM_167075.2 | NP_727096.1 | 2 - 3 | 186 | 73 | ||
| GenBank mRNA | AY089570.1 | - | 4 - 5 | 276 | 73 | |
More Information
Set Membership: |
|

RpL7A
Genomic Map