Assay Details
| Gene Symbol: Mcm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078510.3 | NP_511065.1 | 1 | 2730 | 140 | |
| GenBank mRNA | AF124744.1 | - | 1 | 2668 | 140 | |
| BT088975.1 | - | 1 | 2678 | 140 | ||
More Information
Set Membership: |
|

Mcm6
Genomic Map