Assay Details
Gene Symbol: fs(1)h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272392.2 | NP_001259321.1 | 10 - 11 | 6710 | 115 | |
NM_078523.3 | NP_511078.2 | 10 - 11 | 6686 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
