Assay Details
Gene Symbol: ND-75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078528.3 | NP_511083.1 | 2 - 3 | 280 | 68 | |
NM_167152.2 | NP_727255.1 | 1 - 2 | 252 | 68 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
