Assay Details
Gene Symbol: ND-75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078528.3 | NP_511083.1 | 3 - 4 | 614 | 61 | |
NM_167152.2 | NP_727255.1 | 2 - 3 | 586 | 61 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
