Assay Details
Gene Symbol: alpha-Man-Ia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272473.2 | NP_001259402.1 | 2 - 3 | 831 | 72 | |
NM_078550.4 | NP_511105.2 | 1 - 2 | 656 | 72 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
