Assay Details
| Gene Symbol: sesB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298161.1 | NP_001285090.1 | 2 - 3 | 450 | 92 | |
| NM_078554.3 | NP_511109.1 | 2 - 3 | 399 | 92 | ||
| NM_167246.2 | NP_727448.1 | 3 - 4 | 512 | 92 | ||
| NM_167247.2 | NP_727449.1 | 3 - 4 | 518 | 92 | ||
| NM_167248.2 | NP_727450.1 | 2 - 3 | 405 | 92 | ||
| GenBank mRNA | AY060978.1 | - | 2 - 3 | 382 | 92 | |
| AY070894.1 | - | 1082 | 92 | |||
| BT011069.1 | - | 1862 | 92 | |||
| BT120021.1 | - | 3 - 4 | 493 | 92 | ||
More Information
Set Membership: |
|

sesB
Genomic Map