Assay Details
| Gene Symbol: CkIIbeta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014731.2 | NP_001014731.1 | 6 - 7 | 748 | 101 | |
| NM_001258699.1 | NP_001245628.1 | 6 - 7 | 667 | 101 | ||
| NM_078576.5 | NP_511131.3 | 6 - 7 | 702 | 101 | ||
| NM_080762.4 | NP_542940.1 | 6 - 7 | 789 | 101 | ||
| NM_167304.3 | NP_727561.1 | 6 - 7 | 1036 | 101 | ||
| NM_167305.3 | NP_727562.1 | 6 - 7 | 671 | 101 | ||
| GenBank mRNA | AY113447.1 | - | 6 - 7 | 786 | 101 | |
| BT120031.1 | - | 6 - 7 | 1005 | 101 | ||
| M16535.1 | - | 6 - 7 | 653 | 101 | ||
More Information
Set Membership: |
|

CkIIbeta
Genomic Map