Assay Details
| Gene Symbol: LIMK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169262.1 | NP_001162733.1 | 3 - 4 | 2105 | 84 | |
| NM_001169263.1 | NP_001162734.1 | 2 - 3 | 1291 | 84 | ||
| NM_078584.3 | NP_511139.2 | 4 - 5 | 2171 | 84 | ||
| NM_167326.2 | NP_727621.1 | 3 - 4 | 2068 | 84 | ||
| NM_167327.2 | NP_727622.1 | 2 - 3 | 1358 | 84 | ||
| GenBank mRNA | AY119587.1 | - | 3 - 4 | 1957 | 84 | |
| BT133248.1 | - | 2 - 3 | 1315 | 84 | ||
More Information
Set Membership: |
|

LIMK1
Genomic Map