Assay Details
| Gene Symbol: gce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201664.2 | NP_001188593.1 | 6 - 7 | 2422 | 63 | |
| NM_001272635.2 | NP_001259564.1 | 6 - 7 | 1967 | 63 | ||
| NM_001272637.2 | NP_001259566.1 | 6 - 7 | 2479 | 63 | ||
| NM_078605.3 | NP_511160.2 | 6 - 7 | 2883 | 63 | ||
| GenBank mRNA | BT132669.1 | - | 5 - 6 | 2548 | 63 | |
More Information
Set Membership: |
|

gce
Genomic Map