Assay Details
| Gene Symbol: gce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201664.2 | NP_001188593.1 | 9 - 10 | 3601 | 84 | |
| NM_001272635.2 | NP_001259564.1 | 9 - 10 | 3146 | 84 | ||
| NM_001272637.2 | NP_001259566.1 | 9 - 10 | 3658 | 84 | ||
| NM_078605.3 | NP_511160.2 | 9 - 10 | 4062 | 84 | ||
| GenBank mRNA | BT132669.1 | - | 8 - 9 | 3727 | 84 | |
More Information
Set Membership: |
|

gce
Genomic Map