Assay Details
Gene Symbol: exd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272663.2 | NP_001259592.1 | 4 - 5 | 1108 | 63 | |
NM_001272664.1 | NP_001259593.1 | 4 - 5 | 1061 | 63 | ||
NM_078636.5 | NP_523360.1 | 4 - 5 | 1077 | 63 | ||
NM_167478.3 | NP_727923.1 | 4 - 5 | 1038 | 63 | ||
GenBank mRNA | AY061065.1 | - | 4 - 5 | 1106 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
