Assay Details
| Gene Symbol: if | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169306.2 | NP_001162777.1 | 6 - 7 | 847 | 56 | |
| NM_001169307.1 | NP_001162778.1 | 6 - 7 | 982 | 56 | ||
| NM_078654.2 | NP_523378.2 | 6 - 7 | 888 | 56 | ||
| NM_167544.2 | NP_728021.2 | 6 - 7 | 888 | 56 | ||
More Information
Set Membership: |
|

if
Genomic Map