Assay Details
| Gene Symbol: if | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169306.2 | NP_001162777.1 | 8 - 9 | 962 | 73 | |
| NM_001169307.1 | NP_001162778.1 | 8 - 9 | 1097 | 73 | ||
| NM_078654.2 | NP_523378.2 | 8 - 9 | 1003 | 73 | ||
More Information
Set Membership: |
|

if
Genomic Map