Assay Details
| Gene Symbol: RSG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258793.3 | NP_001245722.1 | 5 - 6 | 2121 | 71 | |
| NM_078656.4 | NP_523380.2 | 5 - 6 | 2121 | 71 | ||
| GenBank mRNA | BT021343.1 | - | 2054 | 71 | ||
More Information
Set Membership: |
|

RSG7
Genomic Map