Assay Details
| Gene Symbol: Ada3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272738.2 | NP_001259667.1 | 1 | 2539 | 131 | |
| NM_078671.5 | NP_523395.1 | 1 | 2539 | 131 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ada3| Gene Symbol: Ada3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272738.2 | NP_001259667.1 | 1 | 2539 | 131 | |
| NM_078671.5 | NP_523395.1 | 1 | 2539 | 131 | ||
Set Membership: |
|