Assay Details
Gene Symbol: Tsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298475.1 | NP_001285404.1 | 4 - 5 | 2688 | 86 | |
NM_078677.3 | NP_523401.2 | 4 - 5 | 1851 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
