Assay Details
| Gene Symbol: pcm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169325.2 | NP_001162796.1 | 10 - 11 | 4955 | 56 | |
| NM_078684.4 | NP_523408.2 | 10 - 11 | 4952 | 56 | ||
| GenBank mRNA | AY051675.1 | - | 10 - 11 | 4892 | 56 | |
More Information
Set Membership: |
|

pcm
Genomic Map