Assay Details
Gene Symbol: smo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078719.4 | NP_523443.1 | 1 - 2 | 285 | 62 | |
GenBank mRNA | BT053691.1 | - | 1 - 2 | 257 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
