Assay Details
| Gene Symbol: Gr21a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272912.1 | NP_001259841.1 | 2 - 3 | 832 | 87 | |
| NM_078724.6 | NP_523448.2 | 2 - 3 | 1626 | 87 | ||
More Information
Set Membership: |
|

Gr21a
Genomic Map