Assay Details
| Gene Symbol: ast | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078727.3 | NP_523451.2 | 1 - 2 | 622 | 60 | |
| GenBank mRNA | BT044345.1 | - | 1 - 2 | 590 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ast| Gene Symbol: ast | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078727.3 | NP_523451.2 | 1 - 2 | 622 | 60 | |
| GenBank mRNA | BT044345.1 | - | 1 - 2 | 590 | 60 | |
Set Membership: |
|