Assay Details
| Gene Symbol: aop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272979.1 | NP_001259908.1 | 4 - 5 | 3013 | 123 | |
| NM_001272980.1 | NP_001259909.1 | 3 - 4 | 2566 | 123 | ||
| NM_001272981.1 | NP_001259910.1 | 3 - 4 | 2956 | 123 | ||
| NM_078731.3 | NP_523455.2 | 3 - 4 | 2956 | 123 | ||
| NM_164457.2 | NP_722766.1 | 3 - 4 | 2343 | 123 | ||
| GenBank mRNA | BT015272.1 | - | 3 - 4 | 2903 | 123 | |
More Information
Set Membership: |
|

aop
Genomic Map