Assay Details
Gene Symbol: RpS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273041.1 | NP_001259970.1 | 2 - 3 | 128 | 65 | |
NM_078738.5 | NP_523462.2 | 2 - 3 | 169 | 65 | ||
NM_164506.4 | NP_722853.1 | 2 - 3 | 128 | 65 | ||
NM_164507.4 | NP_722854.1 | 2 - 3 | 128 | 65 | ||
NM_164508.3 | NP_722855.1 | 2 - 3 | 152 | 65 | ||
GenBank mRNA | BT015189.1 | - | 2 - 3 | 127 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
