Assay Details
Gene Symbol: lilli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144300.2 | NP_001137772.1 | 9 - 10 | 4800 | 86 | |
NM_001144301.1 | NP_001137773.1 | 10 - 11 | 5140 | 86 | ||
NM_001144302.2 | NP_001137774.1 | 9 - 10 | 4909 | 86 | ||
NM_001144303.1 | NP_001137775.1 | 11 - 12 | 6444 | 86 | ||
NM_001273044.1 | NP_001259973.1 | 10 - 11 | 6597 | 86 | ||
NM_001347818.1 | NP_001334729.1 | 10 - 11 | 6597 | 86 | ||
NM_078740.3 | NP_523464.1 | 11 - 12 | 7360 | 86 | ||
NM_164516.2 | NP_722863.1 | 12 - 13 | 7528 | 86 | ||
NM_164517.1 | NP_722864.1 | 11 - 12 | 6249 | 86 | ||
GenBank mRNA | AF289034.1 | - | 12 - 13 | 7505 | 86 | |
AF293971.1 | - | 11 - 12 | 7334 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
