Assay Details
Gene Symbol: lilli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144300.2 | NP_001137772.1 | 2 - 3 | 823 | 82 | |
NM_001144301.1 | NP_001137773.1 | 3 - 4 | 1163 | 82 | ||
NM_001144302.2 | NP_001137774.1 | 2 - 3 | 932 | 82 | ||
NM_001144303.1 | NP_001137775.1 | 4 - 5 | 2467 | 82 | ||
NM_001273044.1 | NP_001259973.1 | 3 - 4 | 2620 | 82 | ||
NM_001347818.1 | NP_001334729.1 | 3 - 4 | 2620 | 82 | ||
NM_078740.3 | NP_523464.1 | 4 - 5 | 3383 | 82 | ||
NM_164516.2 | NP_722863.1 | 5 - 6 | 3551 | 82 | ||
NM_164517.1 | NP_722864.1 | 4 - 5 | 2272 | 82 | ||
GenBank mRNA | AF289034.1 | - | 5 - 6 | 3528 | 82 | |
AF293971.1 | - | 4 - 5 | 3357 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
