Assay Details
Gene Symbol: lilli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144300.2 | NP_001137772.1 | 4 - 5 | 1304 | 68 | |
NM_001144301.1 | NP_001137773.1 | 5 - 6 | 1644 | 68 | ||
NM_001144302.2 | NP_001137774.1 | 4 - 5 | 1413 | 68 | ||
NM_001144303.1 | NP_001137775.1 | 6 - 7 | 2948 | 68 | ||
NM_001273044.1 | NP_001259973.1 | 5 - 6 | 3101 | 68 | ||
NM_001347818.1 | NP_001334729.1 | 5 - 6 | 3101 | 68 | ||
NM_078740.3 | NP_523464.1 | 6 - 7 | 3864 | 68 | ||
NM_164516.2 | NP_722863.1 | 7 - 8 | 4032 | 68 | ||
NM_164517.1 | NP_722864.1 | 6 - 7 | 2753 | 68 | ||
GenBank mRNA | AF289034.1 | - | 7 - 8 | 4009 | 68 | |
AF293971.1 | - | 6 - 7 | 3838 | 68 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
