Assay Details
Gene Symbol: tim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014463.4 | NP_001014463.2 | 11 - 12 | 3068 | 95 | |
NM_001038783.3 | NP_001033872.2 | 10 - 11 | 3495 | 95 | ||
NM_001144308.3 | NP_001137780.2 | 11 - 12 | 3111 | 95 | ||
NM_001347756.1 | NP_001334730.1 | 10 - 11 | 3164 | 95 | ||
NM_164540.4 | NP_722912.3 | 10 - 11 | 3495 | 95 | ||
NM_164541.6 | NP_722913.3 | 10 - 11 | 3168 | 95 | ||
NM_164542.5 | NP_722914.3 | 10 - 11 | 3164 | 95 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
