Assay Details
| Gene Symbol: tim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014463.4 | NP_001014463.2 | 12 - 13 | 3227 | 66 | |
| NM_001038783.3 | NP_001033872.2 | 11 - 12 | 3654 | 66 | ||
| NM_001144308.3 | NP_001137780.2 | 12 - 13 | 3270 | 66 | ||
| NM_001347756.1 | NP_001334730.1 | 11 - 12 | 3323 | 66 | ||
| NM_164540.4 | NP_722912.3 | 11 - 12 | 3654 | 66 | ||
| NM_164541.6 | NP_722913.3 | 11 - 12 | 3327 | 66 | ||
| NM_164542.5 | NP_722914.3 | 11 - 12 | 3323 | 66 | ||
More Information
Set Membership: |
|

tim
Genomic Map