Assay Details
Gene Symbol: tim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014463.4 | NP_001014463.2 | 6 - 7 | 1939 | 100 | |
NM_001038783.3 | NP_001033872.2 | 5 - 6 | 2366 | 100 | ||
NM_001144308.3 | NP_001137780.2 | 6 - 7 | 1982 | 100 | ||
NM_001144309.4 | NP_001137781.2 | 5 - 6 | 2421 | 100 | ||
NM_001201757.2 | NP_001188686.2 | 5 - 6 | 2035 | 100 | ||
NM_001347756.1 | NP_001334730.1 | 5 - 6 | 2035 | 100 | ||
NM_164540.4 | NP_722912.3 | 5 - 6 | 2366 | 100 | ||
NM_164541.6 | NP_722913.3 | 5 - 6 | 2042 | 100 | ||
NM_164542.5 | NP_722914.3 | 5 - 6 | 2035 | 100 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
