Assay Details
| Gene Symbol: tim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014463.4 | NP_001014463.2 | 7 - 8 | 2082 | 61 | |
| NM_001038783.3 | NP_001033872.2 | 6 - 7 | 2509 | 61 | ||
| NM_001144308.3 | NP_001137780.2 | 7 - 8 | 2125 | 61 | ||
| NM_001144309.4 | NP_001137781.2 | 6 - 7 | 2564 | 61 | ||
| NM_001347756.1 | NP_001334730.1 | 6 - 7 | 2178 | 61 | ||
| NM_164540.4 | NP_722912.3 | 6 - 7 | 2509 | 61 | ||
| NM_164542.5 | NP_722914.3 | 6 - 7 | 2178 | 61 | ||
More Information
Set Membership: |
|

tim
Genomic Map