Assay Details
| Gene Symbol: mRpL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078752.3 | NP_523476.1 | 2 - 3 | 325 | 56 | |
| GenBank mRNA | AY095024.1 | - | 2 - 3 | 301 | 56 | |
More Information
Set Membership: |
|

mRpL24
Genomic Map