Assay Details
| Gene Symbol: eIF3i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078754.4 | NP_523478.1 | 2 - 3 | 581 | 90 | |
| GenBank mRNA | AY118527.1 | - | 2 - 3 | 560 | 90 | |
| U90930.1 | - | 2 - 3 | 556 | 90 | ||
More Information
Set Membership: |
|

eIF3i
Genomic Map