Assay Details
| Gene Symbol: GluRIIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078760.3 | NP_523484.2 | 9 - 10 | 1960 | 73 | |
| GenBank mRNA | BT010006.1 | - | 9 - 10 | 1958 | 73 | |
More Information
Set Membership: |
|

GluRIIA
Genomic Map