Assay Details
Gene Symbol: mtm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298722.1 | NP_001285651.1 | 1 - 2 | 1742 | 75 | |
NM_078765.4 | NP_523489.1 | 1 - 2 | 1644 | 75 | ||
GenBank mRNA | AY058594.1 | - | 1 - 2 | 1623 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
