Assay Details
Gene Symbol: cort | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273215.1 | NP_001260144.1 | 1 - 2 | 140 | 118 | |
NM_078770.2 | NP_523494.1 | 1 - 2 | 208 | 118 | ||
GenBank mRNA | AY033478.1 | - | 1 - 2 | 208 | 118 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
