Assay Details
Gene Symbol: cort | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273215.1 | NP_001260144.1 | 2 - 3 | 276 | 64 | |
NM_078770.2 | NP_523494.1 | 2 - 3 | 344 | 64 | ||
GenBank mRNA | AY033478.1 | - | 2 - 3 | 344 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
