Assay Details
Gene Symbol: Gart | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014477.2 | NP_001014477.1 | 1 - 2 | 368 | 70 | |
NM_001298769.1 | NP_001285698.1 | 1 - 2 | 368 | 70 | ||
NM_078773.3 | NP_523497.2 | 1 - 2 | 368 | 70 | ||
GenBank mRNA | BT030993.1 | - | 1 - 2 | 179 | 70 | |
BT031204.1 | - | 1 - 2 | 348 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
