Assay Details
| Gene Symbol: AsnRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273632.1 | NP_001260561.1 | 1 - 2 | 160 | 74 | |
| NM_136104.4 | NP_609948.1 | 1 - 2 | 160 | 74 | ||
| GenBank mRNA | AF181634.1 | - | 1 - 2 | 142 | 74 | |
More Information
Set Membership: |
|

AsnRS
Genomic Map