Assay Details
Gene Symbol: Taf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136180.4 | NP_610024.1 | 1 - 2 | 132 | 77 | |
GenBank mRNA | AY071439.1 | - | 1 - 2 | 129 | 77 | |
KX531312.1 | - | 1 - 2 | 51 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
