Assay Details
| Gene Symbol: RPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273723.1 | NP_001260652.1 | 2 - 3 | 176 | 65 | |
| NM_136233.3 | NP_610077.1 | 2 - 3 | 176 | 65 | ||
| GenBank mRNA | AY069259.1 | - | 2 - 3 | 151 | 65 | |
More Information
Set Membership: |
|

RPA2
Genomic Map