Assay Details
| Gene Symbol: Nipped-A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103722.3 | NP_001097192.2 | 6 - 7 | 1071 | 123 | |
| NM_001316406.1 | NP_001303335.1 | 6 - 7 | 1071 | 123 | ||
| GenBank mRNA | DQ352451.1 | - | 6 - 7 | 1097 | 123 | |
More Information
Set Membership: |
|

Nipped-A
Genomic Map