Assay Details
Gene Symbol: Nipped-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103722.3 | NP_001097192.2 | 13 - 14 | 4180 | 109 | |
NM_001316406.1 | NP_001303335.1 | 13 - 14 | 4180 | 109 | ||
GenBank mRNA | DQ352451.1 | - | 13 - 14 | 4206 | 109 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
