Assay Details
Gene Symbol: TpnC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169584.2 | NP_001163055.1 | 4 - 5 | 513 | 124 | |
NM_136329.3 | NP_610173.2 | 3 - 4 | 616 | 124 | ||
GenBank mRNA | AY283602.1 | - | 4 - 5 | 510 | 124 | |
BT044320.1 | - | 3 - 4 | 627 | 124 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
