Assay Details
| Gene Symbol: TpnC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169584.2 | NP_001163055.1 | 5 - 6 | 527 | 83 | |
| NM_136329.3 | NP_610173.2 | 4 - 5 | 630 | 83 | ||
| GenBank mRNA | AY283602.1 | - | 5 - 6 | 524 | 83 | |
| BT044320.1 | - | 4 - 5 | 641 | 83 | ||
More Information
Set Membership: |
|

TpnC4
Genomic Map