Assay Details
| Gene Symbol: Mob4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259222.2 | NP_001246151.1 | 1 - 2 | 323 | 96 | |
| NM_136385.4 | NP_610229.1 | 1 - 2 | 323 | 96 | ||
| GenBank mRNA | AY070538.1 | - | 1 - 2 | 266 | 96 | |
More Information
Set Membership: |
|

Mob4
Genomic Map