Assay Details
| Gene Symbol: Mob4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136385.4 | NP_610229.1 | 2 - 3 | 699 | 72 | |
| GenBank mRNA | AY070538.1 | - | 2 - 3 | 642 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mob4| Gene Symbol: Mob4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136385.4 | NP_610229.1 | 2 - 3 | 699 | 72 | |
| GenBank mRNA | AY070538.1 | - | 2 - 3 | 642 | 72 | |
Set Membership: |
|